missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Aphidicolin

A DNA polymerase inhibitor, blocks the cell cycle at the early S-phase

151.76 € - 557.78 €

Chemical Identifiers

CAS 38966-21-1
Molecular Formula C20H34O4
Synonym APC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15453039
View Documents
Thermo Scientific Chemicals
J60236.M^
1 mg
151.76 €
1mg
Please to purchase this item. Need a web account? Register with us today!
15443039
View Documents
Thermo Scientific Chemicals
J60236.#0
5 mg
557.78 €
5mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A DNA polymerase inhibitor. Blocks the cell cycle at the early S-phase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

38966-21-1
C20H34O4
APC

Specifications

White
A DNA polymerase inhibitor. Blocks the cell cycle at the early S-phase
C20H34O4
APC
Soluble in DMSO or methanol. Insoluble in water. Store solutions at -20°
Cyrstalline powder
NOFOAYPPHIUXJR-APNQCZIXSA-N
(1S,2S,5R,6R,7R,10S,12R,13R)-6,13-bis(hydroxymethyl)-2,6-dimethyltetracyclo[10.3.1.0¹,¹⁰.0²,⁷]hexadecane-5,13-diol
2°C to 8°C
38966-21-1
338.48
224°C
Aphidicolin
4689958
C[C@@]1(CO)[C@H](O)CC[C@@]2(C)[C@H]1CC[C@H]1C[C@@H]3C[C@]21CC[C@]3(O)CO
338.49
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.