Learn More
Invitrogen™ Anza™ 36 Eco31I
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Brand:  Invitrogen™ IVGN0366
Product Code. 15431933
  • 107.00 € / 800 units
Explore more special offers
Includes: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Description
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
  • One buffer for all restriction enzymes
  • One digestion protocol for all DNA types
  • Complete digestion in 15 min
  • Overnight digestion without star activity
Certifications
CoA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
80 μL Anza 36 Eco31I
500 μL Anza Buffer (10X)
500 μL Anza Red Buffer (10X)
Store at -20°C.
CpG Methylation-Sensitive, Not CpG Methylation-Sensitive, Not Dam Methylation-Sensitive
Anza™ 10X Red Buffer, Anza™ 10X Buffer
37°C
800 units
Traditional Cloning
Approved for shipment on Wet or Dry Ice
Eco31I
Yes
Yes
Restriction Enzyme Cloning System
Anza™