missing translation for 'onlineSavingsMsg'
Learn More

Antimony(V) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11383837
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as a polymerization catalyst, a chlorinating agent, an intermediate in chemical synthesis, also a strong oxidizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-18-9
Molecular Formula Cl5Sb
Molecular Weight (g/mol) 299.02
MDL Number MFCD00011213
InChI Key VMPVEPPRYRXYNP-UHFFFAOYSA-I
Synonym antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
PubChem CID 24294
IUPAC Name antimony pentachloride
SMILES Cl[Sb](Cl)(Cl)(Cl)Cl

Specifications

Boiling Point 79°C (22mmHg)
Melting Point 2.8°C
Physical Form Liquid
Quantity 10 g
UN Number UN1730
Merck Index 14,695
Formula Weight 299.02
Percent Purity 99.999%
Odor Pungent
Refractive Index 1.601
Sensitivity Moisture Sensitive
Density 2.336g/mL
Assay (metals basis)
Chemical Name or Material Antimony(V) chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'