missing translation for 'onlineSavingsMsg'
Learn More

Antimony tribromide, 97%, pure, Thermo Scientific Chemicals

110.55 € - 391.95 €

Chemical Identifiers

CAS 7789-61-9
Molecular Formula Br3Sb
Molecular Weight (g/mol) 361.48
InChI Key RPJGYLSSECYURW-UHFFFAOYSA-K
Synonym antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3
PubChem CID 24615
IUPAC Name tribromostibane
SMILES Br[Sb](Br)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15249826
View Documents
Thermo Scientific Acros
318400500
50 g
110.55 €
50g
Please to purchase this item. Need a web account? Register with us today!
15259826
View Documents
Thermo Scientific Acros
318402500
250 g
391.95 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-61-9
361.48
antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3
tribromostibane
Br3Sb
RPJGYLSSECYURW-UHFFFAOYSA-K
24615
Br[Sb](Br)Br

Specifications

280.0°C
7789-61-9
Br3Sb
15,695
RPJGYLSSECYURW-UHFFFAOYSA-K
tribromostibane
24615
97%
Glass bottle
97.0°C
96% min. (Argentometry)
SbBr3
antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3
Br[Sb](Br)Br
361.48
361.48
Pure
Antimony tribromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
Dispose of contents/container to an approve

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 232-179-8

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.