missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) selenide, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11322228
25 g
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used to prepare large zintl ion, it is now actively explored for thin film solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1315-05-5
MDL Number MFCD00016319

Specifications

Melting Point 611°C
Physical Form Pieces
Quantity 25 g
UN Number UN3077
Merck Index 14,712
Solubility Information Very slightly soluble in water
Formula Weight 480.38
Percent Purity 99.999%
Sensitivity Moisture sensitive
Assay (metals basis)
Chemical Name or Material Antimony(III) selenide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Antimony(III) selenide, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.