Learn More
Antimony(III) chloride, 99.9% (metals basis), Thermo Scientific Chemicals
61.90 € - 173.00 €
Chemical Identifiers
CAS | 10025-91-9 |
---|---|
Molecular Formula | Cl3Sb |
Molecular Weight (g/mol) | 228.11 |
MDL Number | MFCD00011212 |
InChI Key | FAPDDOBMIUGHIN-UHFFFAOYSA-K |
Synonym | antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony |
PubChem CID | 24814 |
ChEBI | CHEBI:74856 |
IUPAC Name | trichlorostibane |
SMILES | [Cl-].[Cl-].[Cl-].[Sb+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11374288
|
Thermo Scientific Alfa Aesar
041537.14 |
25 g |
61.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11384288
|
Thermo Scientific Alfa Aesar
041537.22 |
100 g |
173.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Antimony(III) chloride is engaged to detect certain compounds, such as cholesterols. It is a complexing agent used in the colorimetric Carr-Price test for detecting carotenoids including vitamin A. It is employed as a mordant and a fabric flame retardant. It is used as a catalyst for chlorinationn and polymerization reactions. It is used to coat the steel to make bronze, used as a cutting material of antimony salt and as a preservative in medicine. It is used as an analytical reagent for chloral and aromatics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10025-91-9 | |
228.11 | |
FAPDDOBMIUGHIN-UHFFFAOYSA-K | |
24814 | |
trichlorostibane |
Cl3Sb | |
MFCD00011212 | |
antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony | |
CHEBI:74856 | |
[Cl-].[Cl-].[Cl-].[Sb+3] |
Specifications
223.5°C | |
10025-91-9 | |
Crystals | |
Cl3Sb | |
antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony | |
FAPDDOBMIUGHIN-UHFFFAOYSA-K | |
trichlorostibane | |
24814 | |
228.11 | |
Moisture sensitive | |
Antimony(III) chloride |
73.4°C | |
White | |
99.99% (Metals basis) | |
MFCD00011212 | |
Soluble in water,hydrochloric acid,alcohol,benzene,carbon disulfide,dioxane,chloroform,ether,acetone and carbon tetrachloride. | |
[Cl-].[Cl-].[Cl-].[Sb+3] | |
228.11 | |
CHEBI:74856 | |
Pungent | |
3.14 |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: II
missing translation for 'einecsNumber' : 233-047-2
missing translation for 'rtecsNumber' : CC4900000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only