Learn More
Antimony(III) bromide, 99.995% (metals basis), Thermo Scientific Chemicals
113.00 € - 464.00 €
Chemical Identifiers
CAS | 7789-61-9 |
---|---|
Molecular Formula | Br3Sb |
Molecular Weight (g/mol) | 361.472 |
MDL Number | MFCD00016317 |
InChI Key | RPJGYLSSECYURW-UHFFFAOYSA-K |
Synonym | antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3 |
PubChem CID | 24615 |
IUPAC Name | tribromostibane |
SMILES | Br[Sb](Br)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11301798
|
Thermo Scientific Alfa Aesar
011686.09 |
10 g |
113.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11311798
|
Thermo Scientific Alfa Aesar
011686.18 |
50 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-61-9 | |
361.472 | |
RPJGYLSSECYURW-UHFFFAOYSA-K | |
24615 | |
Br[Sb](Br)Br |
Br3Sb | |
MFCD00016317 | |
antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3 | |
tribromostibane |
Specifications
280°C | |
7789-61-9 | |
Br3Sb | |
UN3260 | |
antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3 | |
Br[Sb](Br)Br | |
361.472 | |
361.48 | |
Characteristic | |
Moisture sensitive | |
(metals basis) |
96.6°C | |
Crystalline | |
MFCD00016317 | |
14,706 | |
RPJGYLSSECYURW-UHFFFAOYSA-K | |
tribromostibane | |
24615 | |
99.995% | |
1.74 | |
4.148 g/mL | |
Antimony(III) bromide |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 232-179-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only