missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Anticon 100™ StandardWeight Wipes
BIOPHARMA

Anticon 100™ StandardWeight wipes are made from 100% continuous filament polyester with a double-knit no-run interlock construction.

304.00 € - 559.00 €

Specifications

Product Type Cleanroom Wipe
Material Polyester
Autoclavable Not Autoclavable
Description Flat Stacked
Disposable Single-use
View More Specs

Products 2
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
16665504
BIOPHARMA
View Documents
Contec™
495352-803
23 x 23 cm (9 x 9 in.)
559.00 €
Pack of 1200
Estimated Shipment: 11-02-2025
to see stock.
Add to basket
 
16675504
BIOPHARMA
View Documents
Contec™
495352-805
30 x 30 cm (12 x 12 in.)
304.00 €
Pack of 400
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
 
Description

Description

The clean cut edges create a very soft wipe, where the whole of the wipe fabric can be used with no risk of scratching. The wipes are cleanroom laundered and packed in a class 4 cleanroom and exhibit very low particle and fibre counts for a knife cut wipe helping to minimize contamination. Using patented textile technology Anticon 100 Standard Weight wipes achieve high capacity with a quick sorption rate. Ideal for spill control as well as applying disinfectants and cleaning solutions.

Features & Benefits

  • Recommended for ISO Class 5-8 environments
  • Manufactured from 100% continuous filament polyester in a no-run interlock knit construction
  • Patented textile technology creates a polyester wipe with high sorbency
  • Laundered and packed in a Class 4 cleanroom, with knife cut edge
  • Lowest fibres and particles available for a cut edge wipe reduces risk of contamination
  • High sorptive capacity decreases the number of wipes required making it an excellent choice for wiping and absorbing spills in critical cleanroom environments
Specifications

Specifications

Cleanroom Wipe
Not Autoclavable
Single-use
Non-sterile
Polyester
Flat Stacked
1-ply
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.