missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR41 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA524352
This item is not returnable.
View return policy
Description
WDR41 Polyclonal antibody specifically detects WDR41 in Human, Mouse samples. It is validated for Western Blot.Specifications
WDR41 | |
Polyclonal | |
Unconjugated | |
WDR41 | |
B830029I03Rik; MSTP048; WD repeat domain 41; WD repeat-containing protein 41; WDR41 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
218460, 55255 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q3UDP0, Q9HAD4 | |
WDR41 | |
KLH conjugated synthetic peptide between 249-277 amino acids from the C-terminal region of human WDR41 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction