missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VWF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA516634
This item is not returnable.
View return policy
Description
PA5-16634 targets von Willebrand Factor in IF and IHC (P) applications and shows reactivity with Human samples. The PA5-16634 immunogen is purified factor VIII related antigen/von Willebrand factor.
Von Willebrand factor (vWf) is a series of self-aggregated structures, all derived from common glycoprotein subunits, synthesized in endothelial cells and megakaryocytes. The molecular mass of the monomeric vWf unit is 225 kDa. vWf binds to circulating factor VIIIc (antihemophilic factor), hereby stabilizing the latter thus being an important factor in the normal primary haemostasis.Specifications
VWF | |
Polyclonal | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P04275, Q8CIZ8 | |
VWF | |
Purified factor VIII related antigen/von Willebrand factor | |
500 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
VWF | |
6820430P06Rik; AI551257; B130011O06Rik; C630030D09; Coagulation Factor VIII; coagulation factor VIII VWF; EMBL:CAB37852.1}; F8VWF; factor viii related antigen; von Willebrand antigen 2; von Willebrand antigen II; von Willebrand Disease (vWD); von Willebrand factor; Von Willebrand factor homolog; VWD; vwf; vwf {ECO:0000312 | |
Rabbit | |
purified | |
RUO | |
22371, 7450 | |
4° C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction