missing translation for 'onlineSavingsMsg'
Learn More
Learn More
regulator of G-protein signaling 5, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant RGS5.
Brand: Abnova H00008490-A01
This item is not returnable.
View return policy
Description
The regulator of G protein signaling (RGS) proteins are signal transduction molecules that have structural homology to SST2 of Saccharomyces cerevisiae and EGL-10 of Caenorhabditis elegans. Multiple genes homologous to SST2 are present in higher eukaryotes. RGS proteins are involved in the regulation of heterotrimeric G proteins by acting as GTPase activators.[supplied by OMIM
Sequence: WIACEDYKKIKSPAKMAEKAKQIYEEFIQTEAPKEVNIDHFTKDITMKNLVEPSLSSFDMAQKRIHALMEKDSLPRFVRSEFYQELIKSpecifications
regulator of G-protein signaling 5 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant RGS5. | |
RGS5 | |
MST092/MST106/MST129/MSTP032/MSTP092/MSTP106/MSTP129 | |
Mouse | |
50 μL | |
Signal Transduction | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_003617 | |
RGS5 | |
RGS5 (NM_003617, 94 a.a. ∼ 181 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
8490 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |