missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB29 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531007
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, Raji. Predicted reactivity: Mouse (91%), Rat (93%), Rhesus Monkey (97%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Rab GTPase key regulator in vesicle trafficking. Essential for maintaining the integrity of the endosome-trans-Golgi network structure. Together with LRRK2, plays a role in the retrograde trafficking pathway for recycling proteins, such as mannose 6 phosphate receptor (M6PR), between lysosomes and the Golgi apparatus in a retromer-dependent manner. Regulates neuronal process morphology in the intact central nervous system (CNS). May play a role in the formation of typhoid toxin transport intermediates during Salmonella enterica serovar Typhi (S.Typhi) epithelial cell infection. {ECO:0000269; PubMed:22042847, ECO:0000269; PubMed:24788816}.Specifications
RAB29 | |
Polyclonal | |
Unconjugated | |
RAB29 | |
RAB29; RAB29, member RAS oncogene family; RAB7, member RAS oncogene family-like 1; RAB7L; RAB7L1; Rab-7-like protein 1; Ras-related GTP-binding protein Rab29; ras-related protein Rab-29; Ras-related protein Rab-7L1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8934 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O14966 | |
RAB29 | |
Recombinant fragment corresponding to a region within amino acids 3 and 203 of Human RAB7L1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |