Learn More
LMBR1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA526795
Product Code. 13241089
  • 474.00 € / 100µg
Explore more special offers
Description
Description
LMBR1L, also known as LIMR (lipocalin-1-interacting membrane receptor), interacts with lipocalin-1 (Lcn-1), a lipocalin member produced by a number of secretory glands and tissues and known to bind a variety of lipophilic compounds. LMBR1L is a 489-amino acid protein with a predicted molecular mass of 55 kDa and 9 putative transmembrane helices. LMBR1L has been shown to mediate cellular uptake of LMBR1L, thus it is characterized as a endocytic receptor. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LMBR1L
Polyclonal
Unconjugated
LMBR1L
1110013E13Rik; AA415653; D15Ertd735e; KIAA1174; limb development membrane protein 1 like; limb development membrane protein 1-like; limb region 1 homolog-like; limb region 1 like; limb region 1 protein homolog-like; limb region 1-like homolog; limb region 1-like protein -like; LIMR; lipocalin-1 interacting membrane receptor; lipocalin-1-interacting membrane receptor; lipocalin-interacting membrane receptor; Lmbr1l; Protein LMBR1L; RGD1309631; UNQ458/PRO783; Uteroglobin receptor
Rabbit
Antigen affinity chromatography, Protein A
RUO
55716
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q6UX01
LMBR1L
KLH conjugated synthetic peptide between 462-489 amino acids from the C-terminal region of human LMBR1L
400 μL