missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G3BP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539397
This item is not returnable.
View return policy
Description
Specifications
G3BP2 | |
Polyclonal | |
Unconjugated | |
G3BP2 | |
KIAA0660 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23881, 305240, 9908 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9UN86, P97379 | |
G3BP2 | |
A synthetic peptide derived from the C-terminal region of human G3BP2 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |