Learn More
FOXN4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA539174
Product Code. 15586081
  • 451.00 € / 100µg
Explore more special offers
Description
Description
Members of the winged-helix/forkhead family of transcription factors, such as FOXN4, are characterized by a 110-amino acid DNA-binding domain that can fold into a variant of the helix-turn-helix motif consisting of 3 alpha helices flanked by 2 large loops or wings. These transcription factors are involved in a variety of biologic processes as key regulators in development and metabolism (Li et al., 2004 [PubMed 15363391]).[supplied by OMIM, Mar 2008].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FOXN4
Polyclonal
Unconjugated
FOXN4
sli, slipjig
Rabbit
Antigen affinity chromatography
RUO
121643
-20°C
Liquid
Western Blot
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Q96NZ1
FOXN4
A synthetic peptide derived from the internal region of human FOXN4