Learn More
CXCL9 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand:  Invitrogen PA547020
Product Code. 15547512
  • 530.00 € / 100µg
Explore more special offers
Description
Description
In sandwich ELISAs, less than 0.5% cross-reactivity with recombinant human (rh) CXCL9/MIG is observed and less than 0.005% cross-reactivity with recombinant mouse MIP-2, recombinant rat CINC-1, rhGRO alpha, rhGRO beta, and rhGRO gamma is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
CXCL9 Polyclonal antibody specifically detects CXCL9 in Mouse samples. It is validated for ELISA, Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CXCL9
Polyclonal
Unconjugated
CXCL9
BB139920; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 9; CMK; crg-10; CXC; CXC motif chemokine; C-X-C motif chemokine 9; C-X-C motif chemokine ligand 9; CXCL; CXCL9; gamma interferon-induced monokine; gamma-interferon-induced monokine; HuMIG; M119; Mig; monokine induced by gamma interferon; monokine induced by interferon gamma; monokine induced by interferon-gamma; MuMIG; protein m119; SCYB9; small inducible cytokine B subfamily (Cys-X-Cys), member 9; small inducible cytokine B9; small-inducible cytokine B9
Goat
Antigen affinity chromatography
RUO
17329
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
ELISA, Neutralization
0.2 mg/mL
PBS with 5% trehalose and No Preservative
P18340
CXCL9
E. coli-derived recombinant mouse CXCL9/MIG