missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CMTM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539106
This item is not returnable.
View return policy
Description
This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies of signaling molecules. The protein encoded by this gene may play an important role in testicular development. Alternatively spliced transcript variants encoding different isoforms have been identified. Naturally occurring read-through transcription occurs between this locus and the neighboring locus CKLF (chemokine-like factor).[provided by RefSeq, Feb 2011].Specifications
CMTM1 | |
Polyclonal | |
Unconjugated | |
CMTM1 | |
CMTM1 | |
A synthetic peptide derived from the internal region of human CKLF1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8IZ96 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
113540 | |
-20°C | |
Liquid |