missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TECK Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547479
This item is not returnable.
View return policy
Description
In direct ELISAs, less than 1% cross-reactivity with recombinant mouse CCL25/TECK is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
TECK Polyclonal antibody specifically detects TECK in Human samples. It is validated for Neutralization, Western Blot.Specifications
TECK | |
Polyclonal | |
Unconjugated | |
CCL25 | |
A130072A22Rik; AI852536; C-C motif chemokine 25; C-C motif chemokine ligand 25; CCL25; chemokine (C-C motif) ligand 25; chemokine TECK; Ck beta-15; CKb15; Scya25; SCYA25TECKvar; small inducible chemokine 25; small inducible cytokine A25; small inducible cytokine subfamily A (Cys-Cys), member 25; small-inducible cytokine A25; Teck; TECKMGC150327; TECKvar; thymus expressed chemokine; thymus-expressed chemokine | |
Goat | |
Antigen affinity chromatography | |
RUO | |
6370 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O15444 | |
CCL25 | |
E. coli-derived recombinant human CCL25/TECK Gln24-Leu150 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |