missing translation for 'onlineSavingsMsg'
Learn More

Anthraquinone, 98%, Thermo Scientific Chemicals

21.11 € - 99.90 €

Chemical Identifiers

CAS 84-65-1
Molecular Formula C14H8O2
Molecular Weight (g/mol) 208.22
MDL Number MFCD00001188
InChI Key RZVHIXYEVGDQDX-UHFFFAOYSA-N
Synonym anthraquinone, 9,10-anthraquinone, 9,10-anthracenedione, anthradione, hoelite, 9,10-dioxoanthracene, corbit, morkit, 9,10-anthrachinon, anthra-9,10-quinone
PubChem CID 6780
ChEBI CHEBI:40448
SMILES O=C1C2=CC=CC=C2C(=O)C2=CC=CC=C12
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10711734
View Documents
Thermo Scientific Acros
104930500
50 g Plastic bottle
21.11 €
50g
Please to purchase this item. Need a web account? Register with us today!
10644752
View Documents
Thermo Scientific Acros
104932500
250 g Plastic bottle
28.14 €
250g
Please to purchase this item. Need a web account? Register with us today!
10316170
View Documents
Thermo Scientific Acros
104930010
1 kg Plastic bottle
99.90 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

84-65-1
208.22
RZVHIXYEVGDQDX-UHFFFAOYSA-N
6780
O=C1C2=CC=CC=C2C(=O)C2=CC=CC=C12
C14H8O2
MFCD00001188
anthraquinone, 9,10-anthraquinone, 9,10-anthracenedione, anthradione, hoelite, 9,10-dioxoanthracene, corbit, morkit, 9,10-anthrachinon, anthra-9,10-quinone
CHEBI:40448

Specifications

84-65-1
Green-Yellow to Tan
185°C
97.5% min. (HPLC)
MFCD00001188
04,20
anthraquinone, 9,10-anthraquinone, 9,10-anthracenedione, anthradione, hoelite, 9,10-dioxoanthracene, corbit, morkit, 9,10-anthrachinon, anthra-9,10-quinone
RZVHIXYEVGDQDX-UHFFFAOYSA-N
9,10-dihydroanthracene-9,10-dione
6780
208.22
Powder
283.0°C to 288.0°C
379.0°C to 381.0°C
Authentic
C14H8O2
07, 781
15, 679
Solubility in water: insoluble. Other solubilities: soluble 0.44g/100g in alcohol (25°C),2.25 in,boiling alcohol; 0.11g/100g in ether (25°C);,0.61g/100g in chloroform (20°C); 0.26g/100g in,benzene (20°C); 0.30g/100g in toluene (25°
O=C1C2=CC=CC=C2C(=O)C2=CC=CC=C12
208.22
CHEBI:40448
98%
Anthraquinone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
May cause cancer.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 201-549-

Product Identifier
  • Anthraquinone

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1B
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Anthraquinone

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.