Learn More
Anthracene, 99%, Thermo Scientific Chemicals
21.30 € - 2075.00 €
Chemical Identifiers
CAS | 120-12-7 |
---|---|
Molecular Formula | C14H10 |
Molecular Weight (g/mol) | 178.23 |
MDL Number | MFCD00001240 |
InChI Key | MWPLVEDNUUSJAV-UHFFFAOYSA-N |
Synonym | paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767 |
PubChem CID | 8418 |
ChEBI | CHEBI:35298 |
IUPAC Name | anthracene |
SMILES | C1=CC2=CC3=CC=CC=C3C=C2C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10030630
|
Thermo Scientific Acros
104860050 |
5 g | Glass Bottle |
21.30 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10783821
|
Thermo Scientific Acros
104861000 |
100 g | Plastic bottle |
47.50 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10176313
|
Thermo Scientific Acros
104865000 |
500 g | Plastic bottle |
171.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11944451
|
Thermo Scientific Acros
104860025 |
2.5 kg | Plastic bottle |
535.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13428298
|
Thermo Scientific Acros
104860100 |
10 kg | Plastic Drum |
2075.00 €
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
120-12-7 | |
178.23 | |
MWPLVEDNUUSJAV-UHFFFAOYSA-N | |
8418 | |
anthracene |
C14H10 | |
MFCD00001240 | |
paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767 | |
CHEBI:35298 | |
C1=CC2=CC3=CC=CC=C3C=C2C=C1 |
Specifications
120-12-7 | |
Yellow | |
121°C | |
98.5% min. (GC) | |
MFCD00001240 | |
12,290; 15,99; 16,13 | |
paranaphthalene, anthracin, green oil, anthracen, tetra olive n2g, anthracen german, anthrazen, anthracene, pure, bis-alkylamino, ccris 767 | |
MWPLVEDNUUSJAV-UHFFFAOYSA-N | |
anthracene | |
8418 | |
178.23 | |
Crystalline Powder |
215.0°C to 218.0°C | |
340.0°C | |
Authentic | |
C14H10 | |
05,657 | |
15,674 | |
Solubility in water: insoluble. Other solubilities: 1g/67mL alcohol - 1g/70mL methanol, 1g/200mL ether - 1g/62mL benzene, 1g/85mL chloroform - 1g/31mL carbon disulfide, 1g/86mL ccl4 - 1g/125mL toluene | |
C1=CC2=CC3=CC=CC=C3C=C2C=C1 | |
178.23 | |
CHEBI:35298 | |
99% | |
Anthracene, 99% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-371-1
missing translation for 'rtecsNumber' : CA9350000
missing translation for 'tsca' : TSCA
- Anthracene
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only