missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell ActivArmr™ 80-813 Foam-Coated Flame Resistant Gloves
Flame resistant gloves provide Arc Flash Hazard/Risk Category 2 protection against electrical arc flash hazards. Aramid Fiber shell allows for safe handling of sharp objects.
374.00 €
Specifications
ANSI Abrasion Rating | ANSI 3 |
---|---|
Arc Flash | Level 2 |
Certifications/Compliance | NFPA 70E Hazard Risk Category 2 |
Certifications/Compliance (Europe) | EN 407: 412110 |
Coating Material | Neoprene |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
12802253
|
Ansell
80813060 |
6 |
374.00 €
12 pairs |
Estimated Shipment: 13-02-2025
Log in to see stock. |
|||||
12872243
|
Ansell
80813070 |
7 |
374.00 €
12 pairs |
Estimated Shipment: 04-12-2024
Log in to see stock. |
|||||
12822253
|
Ansell
80813080 |
8 |
374.00 €
12 pairs |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
12812253
|
Ansell
80813090 |
9 |
374.00 €
12 pairs |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||||
12832253
|
Ansell
80813100 |
10 |
374.00 €
12 pairs |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
12842253
|
Ansell
80813110 |
11 |
374.00 €
12 pairs |
Estimated Shipment: 13-02-2025
Log in to see stock. |
|||||
Description
- Aramid Fiber shell is inherently flame resistant, and allows for safe handling of sharp objects
- Neoprene foam coating enhances grip in dry, wet, or oily conditions
- Arc rating of 12 cal/cm2 meets NFPA 70E Hazard Risk Category 2
- Ergonomic design is comfortable and flexible
Specifications
ANSI 3 | |
NFPA 70E Hazard Risk Category 2 | |
Neoprene | |
ANSI A4 | |
Aramid Fiber | |
Automotive Assembly, Equipment/Machinery Operation, Mining, and Utility Work | |
ActivArmr™ 80-813 |
Level 2 | |
EN 407: 412110 | |
Green, Black | |
Specific | |
2X42C | |
Knit | |
13 |