missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55364-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to ANKRD11.
ANKRD11 is a member of a novel family of ankyrin repeats containing cofactors (ANCOs) that interact with p160 coactivators to inhibit ligand-dependent transactivation. ANKRD11 encodes a large nuclear protein with five ankyrin repeats, and parts of its sequences have been reported as nasopharyngeal carcinoma susceptibility protein and medulloblastoma antigen. This gene also colocalizes and interacts with histone deacetylases.Specifications
ANKRD11 | |
Polyclonal | |
Unconjugated | |
ANKRD11 | |
ANCO 1, ANCO1, ANKRD11, ankyrin repeat domain 11, LZ16, T13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29123 | |
-20°C | |
Liquid |
Western Blot | |
0.65 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6UB99 | |
ANKRD11 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction