missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anisole, ReagentPlus™, 99%, Honeywell™
Click to view available options
Quantity:
250 mL
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | -37°C |
Color | Colorless |
Density | 0.995g/mL at 25°C |
Boiling Point | 154°C |
Flash Point | 51°C (123.8°F) |
Packaging | Stainless Steel Can |
Linear Formula | CH3OC6H5 |
Refractive Index | n20/D 1.516(lit.) |
Quantity | 20 L |
UN Number | UN2222 |
Show More |
Safety and Handling
Product Identifier
- Anisole
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction