missing translation for 'onlineSavingsMsg'
Learn More

Anisoin, 97%, Thermo Scientific Chemicals

Product Code. 11497144
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Anisoin is a useful pre-chromatographic derivatization fluorogenic reagent for LC (liquid chromatography) analysis of guanidino compounds. It is widely used as photoinitiators in polymerization reactions. It is widely used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing, paper varnish and other surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anisoin is a useful pre-chromatographic derivatization fluorogenic reagent for LC (liquid chromatography) analysis of guanidino compounds. It is widely used as photoinitiators in polymerization reactions. It is widely used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing, paper varnish and other surfaces.

Solubility
Soluble in water, ethanol and acetone.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-52-8
Molecular Formula C16H16O4
Molecular Weight (g/mol) 272.30
MDL Number MFCD00008411
InChI Key LRRQSCPPOIUNGX-UHFFFAOYNA-N
Synonym anisoin, 2-hydroxy-1,2-bis 4-methoxyphenyl ethanone, 4,4'-dimethoxybenzoin, p-anisoin, ethanone, 2-hydroxy-1,2-bis 4-methoxyphenyl, benzoin, 4,4'-dimethoxy, p,p'-dimethoxybenzoin, benzoin,4'-dimethoxy, 4,4'-anisoin, 1,2-bis 4-methoxyphenyl-2-hydroxyethan-1-one
PubChem CID 95415
IUPAC Name 2-hydroxy-1,2-bis(4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C=C1)C(O)C(=O)C1=CC=C(OC)C=C1

Specifications

Melting Point 110°C to 114°C
Quantity 100 g
Beilstein 3211892
Solubility Information Soluble in water,ethanol and acetone.
Formula Weight 272.3
Percent Purity 97%
Chemical Name or Material Anisoin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'