Learn More
Invitrogen™ ANGPTL3 Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EMANGPTL3
Description
The Mouse ANGPTL3 ELISA quantitates Ms ANGPTL3 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms ANGPTL3. Principle of the method The Mouse ANGPLT3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
This gene encodes a member of a family of secreted proteins that function in angiogenesis. The encoded protein, which is expressed predominantly in the liver, is further processed into an N-terminal coiled-coil domain-containing chain and a C-terminal fibrinogen chain. The N-terminal chain is important for lipid metabolism, while the C-terminal chain may be involved in angiogenesis. Mutations in this gene cause familial hypobetalipoproteinemia type 2. [provided by RefSeq, Aug 2015].Specifications
Q9R182 | |
40 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
30924 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Endocrinology | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
40.96-10,000 pg/mL | |
40pg/mL to 10,000pg/mL | |
Biotin | |
Proteases, Peptidases & Inhibitors | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse 6Ckine, Activin A, ADAMTS1, Adiponectin, Angiopoietin-3, ANGTL-4 Artemin, CCL28, CD36, Chordin, CRP, E-Cadherin, Epigen, Epiregulin, Fas, Galectin-7, gp130, Granzyme B, Gremlin, IFN-R1, IL-17B, IL-17B R, IL-22, MIP-1b, MMP-2, MMP-3, MMP-10, PDGF-AA, Persephin, sFRP-3, Shh-N, SLAM, TCK-1, TECK, TGFb1, TNF RI, TNF RII, TPO, TRANCE, TremL1, TWEAK, VEGF-B, VEGF-R2, VCAM-1, VEGF. | |
Mouse | |
Colorimetric Microplate Reader | |
hypl | |
<10% | |
HRP | |
RUO | |
Plasma, 1 μL; Serum, 1 μL; Supernatant, 100 μL | |
RP23-101O16.2, hypl | |
1 hr. 20 min. |