Learn More
Sigma Laborzentrifugen™ Angle Rotors for Sigma™ Series 3 Centrifuges
Use these rotors with maximum load and at maximum speed for up to 50,000 cycles with low maintenance. Sigma™ Angle Rotors for Sigma™ Series 3 Centrifuges are ideal for use in research laboratories.
Description
- Fixed-angle rotors are used for sedimentation with high centrifugal forces of up to and more than 60,000 x g
- Allow shorter sedimentation times in comparison to swing-bucket rotors
- Higher rotational speeds and higher centrifugal forces can be achieved
- Sedimentation distance is also generally shorter than with swing-bucket rotors
- Capacity of the rotor chamber can be better utilized than with swing-bucket rotors
- Rotors can be operated continuously at maximum speed and +4°C in nearly all Sigma centrifuges
Specifications
Angle Rotor |