Learn More
Sigma Laborzentrifugen™ Fixed Angle Rotors for Sigma™ Series 2 Centrifuges
Used for sedimentation with high centrifugal forces typically needed in research laboratories. Sigma Laborzentrifugen™ Fixed Angle Rotors for Sigma™ Series 2 Centrifuges are precision-made from modern, certified high-performance materials with traceable batch numbers.
Specifications
Type Angle Rotor
View More Specs
Description
Description
  • Can be continuously operated at maximum speed and +4°C in nearly all Sigma centrifuges
  • Can be used for very many cycles with maximum load and at maximum speed, in most cases up to 50,000 cycles
  • For use with a variety of bucket and tube sizes
Specifications
Specifications
Angle Rotor