missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMPK gamma 1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10290-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a regulatory subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. This subunit is one of the gamma regulatory subunits of AMPK. Alternatively spliced transcript variants encoding distinct isoforms have been observed.Specifications
AMPK gamma 1 | |
Polyclonal | |
Unconjugated | |
PRKAG1 | |
AMPK gamma1, AMPK subunit gamma 1, AMPKG, PRKAG1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5571 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P54619 | |
PRKAG1 | |
AMPK gamma 1 Fusion Protein Ag0302 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |