missing translation for 'onlineSavingsMsg'
Learn More

Ammonium thiocyanate, 98%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10607401
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.12
MDL Number MFCD00011428
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium salt
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]

Specifications

Melting Point 149.0°C
Color Colorless or White
pH 4.5 to 6.0 (5% soln. at 25°C)
Physical Form Crystals
Quantity 3 kg
Assay Percent Range 98%
Linear Formula NH4SCN
Merck Index 15, 558
Solubility Information Solubility in water: 1600g/L (20°C). Other solubilities: soluble in alcohol, acetone and ammonia, practically insoluble in chcl3 and ethyl acetate
Formula Weight 76.12
Percent Purity 98%
Grade ACS Reagent
Packaging Plastic bottle
Residue after Ignition 0.025% max
Specific Gravity 1.3
Density 1.3000g/mL
Chemical Name or Material Ammonium thiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'