missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium sulphate, granules FCC, J.T. Baker™
Click to view available options
2.5 kg
Description
- Food GMP Manufactured Product
- Meets F.C.C. Requirements
- Bulk Food Chemical
Specifications
Specifications
Melting Point | 336°C to 339°C |
CAS | 7783-20-2 |
Quantity | 2.5 kg |
Assay Percent Range | 99.0 to 100.5% |
Molecular Formula | H8N2O4S |
MDL Number | MFCD00003391 |
Synonym | Diammonium sulphate , Sulfuric acid diammonium salt |
InChI Key | BFNBIHQBYMNNAN-UHFFFAOYSA-N |
SMILES | N.N.OS(O)(=O)=O |
IUPAC Name | sulfuric acid diamine |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'