missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Sulfate, Ultrapure Bioreagent, J.T. Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

135.00 € - 502.00 €

Chemical Identifiers

CAS 7783-20-2
Molecular Formula H8N2O4S
Molecular Weight (g/mol) 132.13
MDL Number MFCD00003391
InChI Key BFNBIHQBYMNNAN-UHFFFAOYSA-N
Synonym ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphateShow MoreShow Less
PubChem CID 6097028
ChEBI CHEBI:62946
IUPAC Name diazanium;sulfate
SMILES N.N.OS(O)(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15538214
View Documents
Avantor J.T.Baker
4027-02
1 kg Poly Bottle
135.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
15311882
View Documents
Avantor J.T.Baker
4027-06
5 kg Poly Pail
502.00 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

    For Protein Precipitation, Liquid Chromatography and Molecular Biology Applications
Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Sulfate, Ultrapure Bioreagent, J.T. Baker™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.