missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfate,≥99%, ReagentPlus™,, Honeywell™
Reagent Grade,≥99.0%
36.54 € - 174.39 €
Chemical Identifiers
CAS | 7783-20-2 |
---|---|
Molecular Formula | H8N2O4S |
Molecular Weight (g/mol) | 132.13 |
MDL Number | MFCD00003391 |
InChI Key | BFNBIHQBYMNNAN-UHFFFAOYSA-N |
Synonym | ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate |
PubChem CID | 6097028 |
ChEBI | CHEBI:62946 |
IUPAC Name | diazanium;sulfate |
SMILES | N.N.OS(O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15657650
|
Honeywell Research Chemicals
A5132-500G |
500 g |
36.54 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15647650
|
Honeywell Research Chemicals
A5132-1KG |
1 kg |
50.67 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15667650
|
Honeywell Research Chemicals
A5132-5KG |
5 kg |
174.39 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7783-20-2 | |
132.13 | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
6097028 | |
diazanium;sulfate |
H8N2O4S | |
MFCD00003391 | |
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
CHEBI:62946 | |
N.N.OS(O)(=O)=O |
Specifications
7783-20-2 | |
(NH4)2SO4 | |
NONH for all modes of transport | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
diazanium;sulfate | |
6097028 | |
132.14g/mol | |
1.77g/mL at 25°C |
H8N2O4S | |
MFCD00003391 | |
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
N.N.OS(O)(=O)=O | |
132.13 | |
CHEBI:62946 | |
≥99% | |
Ammonium sulfate |
Safety and Handling
missing translation for 'rtecsNumber' : BS4500000