Learn More
Ammonium sulfate, 99+%, ACS reagent, Thermo Scientific Chemicals
27.03 € - 629.13 €
Chemical Identifiers
CAS | 7783-20-2 |
---|---|
Molecular Formula | H8N2O4S |
Molecular Weight (g/mol) | 132.13 |
MDL Number | MFCD00003391 |
InChI Key | BFNBIHQBYMNNAN-UHFFFAOYSA-N |
Synonym | ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate |
PubChem CID | 6097028 |
ChEBI | CHEBI:62946 |
SMILES | N.N.OS(O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10556792
|
Thermo Scientific Acros
423400250 |
25 g | Plastic bottle |
27.03 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10697561
|
Thermo Scientific Acros
423405000 |
500 g | Plastic bottle |
33.57 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10556602
|
Thermo Scientific Acros
423400010 |
1 kg | Plastic bottle |
51.36 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10105563
|
Thermo Scientific Acros
423400050 |
5 kg | Plastic bottle |
165.83 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15109293
|
Thermo Scientific Acros
423400251 |
25 kg | Fibre drum |
629.13 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-20-2 | |
132.13 | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
6097028 | |
N.N.OS(O)(=O)=O |
H8N2O4S | |
MFCD00003391 | |
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
CHEBI:62946 |
Specifications
280.0°C | |
Silver to Gray | |
Granules, Fillings or Chips | |
H8N2O4S | |
MFCD00003391 | |
ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
diazanium;sulfate | |
6097028 | |
132.14 | |
ACS Reagent | |
1.76g/mL |
7783-20-2 | |
5.0 to 6.0 (5% soln. at 25°C) | |
99+% | |
(NH4)2SO4 | |
15, 552 | |
Solubility in water: 754g/L. Other solubilities: insoluble in alcohol and acetone | |
N.N.OS(O)(=O)=O | |
132.13 | |
CHEBI:62946 | |
≥99% | |
0.005 % max. | |
Ammonium sulfate |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 231-984-1
- Ammonium sulfate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.