missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Dibasic, puriss. p.a., Honeywell Fluka™

Product Code. 15620920
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a.,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
PubChem CID 24540
ChEBI CHEBI:63051
IUPAC Name diazanium;hydrogen phosphate
SMILES N.N.OP(O)(O)=O

Specifications

Quantity 50 g
Linear Formula (NH4)2HPO4
UN Number NONH for all modes of transport
Formula Weight 132.06g/mol
Percent Purity ≥99% (T)
Grade puriss. p.a.
Chemical Name or Material Ammonium Phosphate Dibasic
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.