Learn More
Ammonium persulfate, 98%, extra pure, Thermo Scientific Chemicals
21.41 € - 308.54 €
Chemical Identifiers
CAS | 7727-54-0 |
---|---|
Molecular Formula | H8N2O8S2 |
Molecular Weight (g/mol) | 228.19 |
MDL Number | MFCD00003390 |
InChI Key | ROOXNKNUYICQNP-UHFFFAOYSA-N |
Synonym | ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french |
PubChem CID | 62648 |
IUPAC Name | diazanium;sulfonatooxy sulfate |
SMILES | [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11316115
|
Thermo Scientific Acros
201531000 |
100 g | Glass bottle |
21.41 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10266400
|
Thermo Scientific Acros
201530010 |
1 kg | Glass bottle |
67.13 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10704304
|
Thermo Scientific Acros
201530050 |
5 kg | Plastic Bottle |
308.54 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
7727-54-0 | |
228.19 | |
ROOXNKNUYICQNP-UHFFFAOYSA-N | |
62648 | |
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O |
H8N2O8S2 | |
MFCD00003390 | |
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french | |
diazanium;sulfonatooxy sulfate |
Specifications
120.0°C | |
White to Yellow | |
98% | |
(NH4)2S2O8 | |
01,952; 02,348; 03,238; 05,15; 06,20; 12,33 | |
ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french | |
ROOXNKNUYICQNP-UHFFFAOYSA-N | |
diazanium;sulfonatooxy sulfate | |
62648 | |
98% | |
1.98 | |
Ammonium persulfate |
7727-54-0 | |
Crystalline Powder and/or Crystals | |
H8N2O8S2 | |
MFCD00003390 | |
15, 539 | |
Solubility in water: 582g/L (20°C), may decompose. | |
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O | |
228.19 | |
228.19 | |
Extra Pure | |
1.9800g/mL |
Safety and Handling
GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Take any precaution to avoid mixing with combustibles.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-786-5
- Ammonium persulfate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.