missing translation for 'onlineSavingsMsg'
Learn More

Ammonium iodide, 98+%, Thermo Scientific Chemicals

Product Code. 11464134
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Ammonium iodide is a laboratory reagent and used as a photographic chemicals. It has wide range of applications in chemical industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium iodide is a laboratory reagent and used as a photographic chemicals. It has wide range of applications in chemical industries.

Solubility
Soluble in water (1770 mg/ml at 25°C), methanol (400 mg/ml), glycerol (667 mg/ml), alcohol (270 mg/ml), and ethanol.

Notes
Store at 4°C. Hande under dry protective gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12027-06-4
Molecular Formula H4IN
Molecular Weight (g/mol) 144.94
MDL Number MFCD00011424
InChI Key UKFWSNCTAHXBQN-UHFFFAOYSA-N
Synonym ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
PubChem CID 25487
ChEBI CHEBI:63040
IUPAC Name azanium;iodide
SMILES [NH4+].[I-]

Specifications

Melting Point 551°C (sublimation)
Quantity 500 g
Merck Index 14,529
Solubility Information Soluble in water (1770mg/ml at 25°C),methanol (400mg/ml),glycerol (667mg/ml),alcohol (270mg/ml),and ethanol.
Formula Weight 144.94
Percent Purity ≥98%
Odor Odorless
Refractive Index 1.7031
Sensitivity Air and light sensitive; Hygroscopic
Density 2.514
Chemical Name or Material Ammonium iodide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'