missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hydroxide solution, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ∼25% NH3 basis, Honeywell Fluka™

Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ∼25% NH3 basis

31.00 € - 498.66 €

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name azanium;hydroxide
SMILES N.O
View More Specs

Products 11
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15653040
SDS View Documents
Honeywell-Fluka
30501-1L-GL
1 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15643040
SDS View Documents
Honeywell-Fluka
30501-1L
1 L Plastic Bottle
38.81 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15673040
SDS View Documents
Honeywell-Fluka
30501-2.5L-GL
2.5 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15663040
SDS View Documents
Honeywell-Fluka
30501-2.5L
2.5 L Plastic Bottle
70.90 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15623050
SDS View Documents
Honeywell-Fluka
30501-5L
5 L Plastic Bottle
141.15 €
5 L
Please to purchase this item. Need a web account? Register with us today!
15643050
SDS View Documents
Honeywell-Fluka
30501-6X1L-GL
6 x 1 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15633050
SDS View Documents
Honeywell-Fluka
30501-6X1L
6 x 1 L Plastic Bottle
211.56 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15693040
SDS View Documents
Honeywell-Fluka
30501-4X2.5L-GL
4 x 2.5 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15683040
SDS View Documents
Honeywell-Fluka
30501-4X2.5L
4 x 2.5 L Plastic Bottle
253.55 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
15603050
SDS View Documents
Honeywell-Fluka
30501-4X5L
4 x 5 L Plastic Bottle
478.10 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
15613050
View Documents
Honeywell-Fluka
30501-50KG
50 kg Metal Drum
498.66 €
50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium hydroxide solution, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ∼25% NH3 basis, Honeywell Fluka™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.