missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide Solution, ≥99.99%, Honeywell™

Product Code. 15684090
Click to view available options
Quantity:
5 mL
100 mL
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

28% NH3 in H2O,≥99.99% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name azanium;hydroxide
SMILES N.O

Specifications

Linear Formula NH4OH
UN Number UN2672
Beilstein 3587154
Density 0.9g/mL at 25°C, 0.9g/mL at 25°C
Quantity 100 mL
Explosion Limit 0.27
Formula Weight 35.05g/mol
Percent Purity ≥99.99% (trace metals basis)
Chemical Name or Material Ammonium hydroxide solution
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium hydroxide solution
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Ammonia, aqueous solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'