missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride, Reagent Grade, ≥99.5%, Honeywell™

Product Code. 15637640
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name azanium;chloride

Specifications

Melting Point 338°C
Color Colorless
pH 4.6 to 6 (at 20°C)
Physical Form Solid
Quantity 100 g
Linear Formula NH4Cl
Beilstein 4371014
SMILES [NH4+].[Cl-]
Formula Weight 53.49g/mol
Percent Purity ≥99.5%
Odor Odorless
Packaging Poly Bottle
Vapor Pressure 1mmHg (160.4°C)
Density 1.536g/cm3 at 20°C; Vapor Density: 1.9 (vs air)
Chemical Name or Material Ammonium chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'