missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbonate, 99.99% Trace Metals Basis, Honeywell™

99.999% trace metals basis

94.63 € - 326.60 €

Chemical Identifiers

CAS 506-87-6
Molecular Formula CH8N2O3
Molecular Weight (g/mol) 96.086
MDL Number MFCD00010890
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PubChem CID 517111
IUPAC Name diazanium;carbonate
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15665180
View Documents Promotion Details
Honeywell Research Chemicals
379999-10G
10 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15675180
View Documents Promotion Details
Honeywell Research Chemicals
379999-50G
50 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications

Chemical Identifiers

506-87-6
96.086
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
517111
C(=O)([O-])[O-].[NH4+].[NH4+]
CH8N2O3
MFCD00010890
ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
diazanium;carbonate

Specifications

506-87-6
9 (at 100g/L 20°C)
(NH4)2CO3
4253487
In water 320g/L at 20°C
C(=O)([O-])[O-].[NH4+].[NH4+]
96.086
96.09g/mol
Ammonia stinging
69 hPa at 20°C
0.9g/cm3 at 20°C; Vapor Density: 2.7 (vs air)
White
CH8N2O3
MFCD00010890
ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
diazanium;carbonate
517111
99.999% (trace metals basis)
Solid
Temperature : >160°C
Ammonium carbonate
Videos
Safety and Handling

Safety and Handling

P301 + P312 + P330

missing translation for 'rtecsNumber' : 208-058-0

SDS
Documents

Documents

Product Certifications