missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Acetate, Honeywell
≥99.99% trace metals basis
132.46 € - 456.29 €
Chemical Identifiers
CAS | 631-61-8 |
---|---|
Molecular Formula | C2H7NO2 |
Molecular Weight (g/mol) | 77.083 |
MDL Number | MFCD00013066 |
InChI Key | USFZMSVCRYTOJT-UHFFFAOYSA-N |
Synonym | ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 |
PubChem CID | 517165 |
ChEBI | CHEBI:62947 |
IUPAC Name | azanium;acetate |
SMILES | CC(=O)[O-].[NH4+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15615510
|
Honeywell Research Chemicals
431311-50G |
50 g |
132.46 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15605510
|
Honeywell Research Chemicals
431311-250G |
250 g |
456.29 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
631-61-8 | |
77.083 | |
USFZMSVCRYTOJT-UHFFFAOYSA-N | |
517165 | |
azanium;acetate |
C2H7NO2 | |
MFCD00013066 | |
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
CHEBI:62947 | |
CC(=O)[O-].[NH4+] |
Specifications
631-61-8 | |
CH3CO2NH4 | |
4186741 | |
USFZMSVCRYTOJT-UHFFFAOYSA-N | |
azanium;acetate | |
517165 | |
77.08g/mol | |
Ammonium acetate |
C2H7NO2 | |
MFCD00013066 | |
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
CC(=O)[O-].[NH4+] | |
77.083 | |
CHEBI:62947 | |
≥99.99% (trace metals basis) |
Safety and Handling
missing translation for 'rtecsNumber' : AF3675000