missing translation for 'onlineSavingsMsg'
Learn More

Ammonium acetate, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10714391
Click to view available options
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Meets ACS specifications

Specifications

Melting Point 114°C
CAS 631-61-8
pH 6.7 to 7.3 (5% Solution at 25°C)
Quantity 500 g
Assay Percent Range min. 97 %
Molecular Formula C2H7NO2
MDL Number MFCD00013066
Flash Point 136°C
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES [NH4+].CC([O-])=O
IUPAC Name ammonium acetate
Molecular Weight (g/mol) 77.08
Packaging Plastic bottle for solids
Residue after Ignition max. 0.01 %
Density 1.17 g/cm³ (20°C)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.