missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia solution 25%, BAKER ANALYZED™, J.T. Baker™, Avantor™
Laboratory Reagent
40.00 € - 80.00 €
Chemical Identifiers
CAS | 1336-21-6 |
---|---|
Molecular Formula | NH3 |
Molecular Weight (g/mol) | 35.05 |
MDL Number | MFCD00066650 |
Synonym | Ammonium hydroxide |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10700312
|
JT Baker
6051.1000 |
1 L |
40.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10691552
|
JT Baker
6051.2500 |
2.5 L |
80.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Chemical Identifiers
1336-21-6 | |
35.05 | |
Ammonium hydroxide |
NH3 | |
MFCD00066650 |
Specifications
32°C to 37.7°C (1013 hPa) | |
-60°C to -57.5°C | |
MFCD00066650 | |
Ammonium hydroxide | |
Glass bottle |
1336-21-6 | |
NH3 | |
0.88 - 0.91 g/cm³ (20°C) | |
35.05 |
Safety and Handling
Recommended Storage : Ambient
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title