missing translation for 'onlineSavingsMsg'
Learn More

Aminoguanidine nitrate, 99%, Thermo Scientific Chemicals

17.59 € - 109.55 €

Chemical Identifiers

CAS 10308-82-4
Molecular Formula CH7N5O3
Molecular Weight (g/mol) 137.10
MDL Number MFCD00013174
InChI Key PMGFHEJUUBDCLU-UHFFFAOYSA-N
Synonym aminoguanidine nitrate, aminoguanidine, nitrate, hydrazinecarboximidamide, mononitrate, monoaminoguanidium nitrate, aminoguanidine, mononitrate, hydrazinecarboximidamide nitrate, hydrazinecarboximidamide, nitrate 1:1, 1-aminoguanidine; nitric acid, aminoguanidine; nitric acid
PubChem CID 165859
IUPAC Name 2-aminoguanidine;nitric acid
SMILES O[N+]([O-])=O.NN=C(N)N
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10254510
View Documents
Thermo Scientific Acros
103760050
5 g Glass bottle
17.59 €
5g
Please to purchase this item. Need a web account? Register with us today!
10560584
View Documents
Thermo Scientific Acros
103760250
25 g Plastic bottle
39.90 €
25g
Please to purchase this item. Need a web account? Register with us today!
10142800
View Documents
Thermo Scientific Acros
103761000
100 g Plastic bottle
109.55 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10308-82-4
137.10
PMGFHEJUUBDCLU-UHFFFAOYSA-N
165859
O[N+]([O-])=O.NN=C(N)N
CH7N5O3
MFCD00013174
aminoguanidine nitrate, aminoguanidine, nitrate, hydrazinecarboximidamide, mononitrate, monoaminoguanidium nitrate, aminoguanidine, mononitrate, hydrazinecarboximidamide nitrate, hydrazinecarboximidamide, nitrate 1:1, 1-aminoguanidine; nitric acid, aminoguanidine; nitric acid
2-aminoguanidine;nitric acid

Specifications

10308-82-4
White
99%
H2NNHC(=NH)NH2·HNO3
03,117
PMGFHEJUUBDCLU-UHFFFAOYSA-N
2-aminoguanidine;nitric acid
165859
99%
Aminoguanidine nitrate, 99%
145°C to 147°C
Authentic
CH7N5O3
MFCD00013174
aminoguanidine nitrate, aminoguanidine, nitrate, hydrazinecarboximidamide, mononitrate, monoaminoguanidium nitrate, aminoguanidine, mononitrate, hydrazinecarboximidamide nitrate, hydrazinecarboximidamide, nitrate 1:1, 1-aminoguanidine; nitric acid, aminoguanidine; nitric acid
O[N+]([O-])=O.NN=C(N)N
137.10
137.11
Fine Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 233-682-5

missing translation for 'tsca' : TSCA

Product Identifier
  • Aminoguanidine nitrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.