missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Twill Jean Wipes
BIOPHARMAGREENER_CHOICE

Contec™ Twill Jean Wipes feature high absorbency and thermal stability, making them ideal for high temperature applications.

Brand:  Contec™ TJ-99

1100.00 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


Product Code. 15855204

  • 1545.00 € / Pack of 3600
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 100% cotton fabric
  • Woven in a 2 x 1 twill construction
  • Strong and absorbent
  • Suitable for high temperature applications
  • Available irradiated
  • Durable fabric for tough tasks
  • Bias cut edges to reduce unraveling
  • Sorbent wipe for surface cleaning or spill pick up
Specifications

Specifications

Wipe
22.86 cm
22.86 cm
Flat stacked
300/Bag, 12 Bags/Cs.
9 in.
9 in.
Cotton
9 x 9 in. (22.86 x 22.86 cm)
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.