missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Formamide (Deionized)

Ambion formamide has been deionized, is nuclease-free, and comes packaged as a liquid in an amber glass bottle under argon.

Brand:  Invitrogen™ AM9342

148.00 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.



 View more versions of this product

Product Code. 10742297

  • 185.00 € / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Ambion formamide has been deionized, is nuclease-free, and comes packaged as a liquid in an amber glass bottle under argon. Supplied in 5 bottles containing 500 g each. Formamide lowers the melting temperature of nucleic acid duplexes and is widely used in applications such as Southern and northern blotting, nuclease protection assays, S1 mapping, and in situ hybridization. For critical applications, it is recommended that the formamide from previously opened bottles be deionized before use.

Appearance: Clear, colorless
Molecular Weight: 45.04
Purity: ≥99.5%
Quality Control: The product has been tested for RNase and DNase contamination.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Colorless
Bottle
Ambion™
500 g
Liquid
Store at 4° or -20°C as small aliquots.
Formamide
>99.5%
Room Temperature
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Formamide (Deionized)

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Formamide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.