missing translation for 'onlineSavingsMsg'
Learn More

Amberlyst™ A26, OH-form, ion-exchange resin, Thermo Scientific Chemicals

59.70 € - 283.41 €

Chemical Identifiers

CAS 39339-85-0
MDL Number MFCD00145579
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10547434
View Documents Promotion Details
Thermo Scientific Acros
202181000
100 mL Plastic bottle
59.70 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10656412
View Documents Promotion Details
Thermo Scientific Acros
202180010
1 L Plastic bottle
283.41 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
Specifications

Chemical Identifiers

39339-85-0
MFCD00145579

Specifications

39339-85-0
0.6750g/mL
10, 381
MFCD00145579
01,511; 02,227; 04,266; 05,355; 06,302; 07,182; 08,263
Amberlyst™ A26
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Amberlyst A26 hydroxide form

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only