missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRC-748, ion exchange resin, Thermo Scientific Chemicals

45.33 € - 126.63 €

Chemical Identifiers

CAS 79620-28-3
MDL Number MFCD00132702
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11315345
View Documents
Thermo Scientific Alfa Aesar
L19570.30
250 g
45.33 €
250g
Please to purchase this item. Need a web account? Register with us today!
11325345
View Documents
Thermo Scientific Alfa Aesar
L19570.0B
1000 g
126.63 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Amberlite™ IRC-748, ion exchange resin is used as a chelating resin with high affinity for heavy metal cations compared to alkali or alkaline earth metals. It is also utilized in metal-chelation applications. Further, it is used in purification of brine in the chloralkali industry as well as the purification of process streams having trace metals. In addition to this, it is used in the electroplating industry and in the recovery of heavy metals from hydrometallurgical leach streams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlite|r IRC-748, ion exchange resin is used as a chelating resin with high affinity for heavy metal cations compared to alkali or alkaline earth metals. It is also utilized in metal-chelation applications. Further, it is used in purification of brine in the chloralkali industry as well as the purification of process streams having trace metals. In addition to this, it is used in the electroplating industry and in the recovery of heavy metals from hydrometallurgical leach streams.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

79620-28-3
MFCD00132702

Specifications

79620-28-3
MFCD00132702
Insoluble in water.
Odorless
14,382
Amberlite™ IRC-748, ion exchange resin
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.