Learn More
Aluminum sulfate, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals
51.15 € - 1816.04 €
Chemical Identifiers
CAS | 10043-01-3 |
---|---|
Molecular Formula | Al2O12S3 |
Molecular Weight (g/mol) | 342.131 |
MDL Number | MFCD00003423 |
InChI Key | DIZPMCHEQGEION-UHFFFAOYSA-H |
Synonym | aluminum sulfate, dialuminum trisulfate, aluminum sulfate 2:3, filter alum, aluminum sulphate, aluminum sulphate, dialuminum sulfate, papermaker's alum, sulfuric acid, aluminum salt 3:2 |
PubChem CID | 24850 |
ChEBI | CHEBI:74768 |
IUPAC Name | dialuminum;trisulfate |
SMILES | [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3] |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
11465330
|
Thermo Scientific Alfa Aesar
044563.06 |
5 g |
51.15 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11475330
|
Thermo Scientific Alfa Aesar
044563.14 |
25 g |
166.83 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11485330
|
Thermo Scientific Alfa Aesar
044563.22 |
100 g |
501.50 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11495330
|
Thermo Scientific Alfa Aesar
044563.36 |
500 g |
1816.04 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Aluminum sulfate acts as a flocculating agent in the purification of drinking water, and in waste water treatment plants. It acts as a mordant in dyeing, printing, textiles and also used in paper manufacturing. It is a waterproofing agent and accelerator in concrete. Further, it is used as a foaming agent in fire fighting foam, photographic film and in photochemicals. It is also used in styptic pencils, pain relief and in dentistry for gingival retraction cords.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Chemische Identifikatoren
10043-01-3 | |
342.131 | |
DIZPMCHEQGEION-UHFFFAOYSA-H | |
24850 | |
dialuminum;trisulfate |
Al2O12S3 | |
MFCD00003423 | |
aluminum sulfate, dialuminum trisulfate, aluminum sulfate 2:3, filter alum, aluminum sulphate, aluminum sulphate, dialuminum sulfate, papermaker's alum, sulfuric acid, aluminum salt 3:2 | |
CHEBI:74768 | |
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3] |
Spezifikation
770°C | |
3.5 | |
Al2O12S3 | |
14,366 | |
Soluble in water. Slightly soluble in alcohol and dilute mineral acids. Insoluble in ethanol. | |
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3] | |
342.131 | |
CHEBI:74768 | |
99.99% | |
2.71 g/mL | |
Aluminum sulfate, Anhydrous |
10043-01-3 | |
Powder | |
MFCD00003423 | |
aluminum sulfate, dialuminum trisulfate, aluminum sulfate 2:3, filter alum, aluminum sulphate, aluminum sulphate, dialuminum sulfate, papermaker's alum, sulfuric acid, aluminum salt 3:2 | |
DIZPMCHEQGEION-UHFFFAOYSA-H | |
dialuminum;trisulfate | |
24850 | |
342.15 | |
Hygroscopic | |
(metals basis) |
Sicherheit und Handhabung
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P280-P305+P351+P338-P310
H318
missing translation for 'einecsNumber' : 233-135-0
missing translation for 'rtecsNumber' : BD1700000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Packaged under Argon
Recommended Storage : Ambient temperatures
- Aluminum sulfate, anhydrous
Signalwort
- Danger
Gefahrenkategorie
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Gefahrenhinweis
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Sicherheitshinweis
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.