missing translation for 'onlineSavingsMsg'
Learn More

Aluminum spheres, 6.35mm (0.25 in.) dia., 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11315969
Click to view available options
Quantity:
100 Pc.
25 Pc.
500 Pc.
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminum
SMILES [Al]

Specifications

Avg. Mol. Wt. or Mol. Wt. Range Alloy 1100, Al:Cu; ≥99:0.12 wt%
Quantity 25 Pc.
Percent Purity 99%
Form Sphere
Assay (metals basis)
Chemical Name or Material Aluminum spheres, 6.35mm (0.25 in.) dia.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'